accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77421-13-7

IUPAC Name: (2-chlorophenyl)methanesulfonyl chloride
Synonyms: (2-chlorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: CHPZYFXSICSCNY-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1CS(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results