accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77470-53-2

IUPAC Name: 4-(chloromethyl)-2-methyl-1,3-thiazole hydrochloride
Synonyms: 4-(chloromethyl)-2-methyl-1,3-thiazole hydrochloride
Molecular Weight (g/mol): 184.08
Molecular Formula: C5H7Cl2NS
InChi Key: YGKDISJLDVGNOR-UHFFFAOYSA-N
SMILES: Cl.CC1=NC(CCl)=CS1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)