accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOHN

CAS RN 77497-74-6

IUPAC Name: tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms: tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Molecular Weight (g/mol): 233.31
Molecular Formula: C14H19NO2
InChi Key: KRSUDBIZXKOKGA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)