accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 775-64-4

CH3CH3HO

CAS RN 775-64-4

IUPAC Name: 2-(adamantan-1-yl)propan-2-ol
Synonyms: 2-(adamantan-1-yl)propan-2-ol
Molecular Weight (g/mol): 194.32
Molecular Formula: C13H22O
InChi Key: WBKAUEBLTWRERU-UHFFFAOYSA-N
SMILES: CC(C)(O)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results