accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 775351-56-9

IUPAC Name: 2-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms: 2-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol): 245.09
Molecular Formula: C13H16BNO3
InChi Key: PUOBFTBTFCPICR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C#N)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results