accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COBOH3CH3CFN

CAS RN 775351-57-0

IUPAC Name: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol): 247.08
Molecular Formula: C13H15BFNO2
InChi Key: RYJOVQGRIOURGT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(F)C(=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)