accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7756-94-7

IUPAC Name: tert-butyl
Synonyms: t-butyl
Molecular Weight (g/mol): 57.12
Molecular Formula: C4H9
InChi Key: IIVWHGMLFGNMOW-UHFFFAOYSA-N
SMILES: C[C](C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results