accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7757-81-5

IUPAC Name: sodium (2E,4E)-hexa-2,4-dienoate
Synonyms: sodium sorbate
Molecular Weight (g/mol): 134.11
Molecular Formula: C6H7NaO2
InChi Key: LROWVYNUWKVTCU-STWYSWDKSA-M
SMILES: [Na+].C\C=C\C=C\C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results