accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKOHPOOO

CAS RN 7758-11-4

IUPAC Name: dipotassium hydrogen phosphate
Synonyms: dipotassium phosphate potassium phosphate, dibasic
Molecular Weight (g/mol): 174.17
Molecular Formula: HK2O4P
InChi Key: ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES: [K+].[K+].OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (16)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
Grade 
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
Physical Form 
  • (13)
  • (2)
Percent Purity 
  • (4)
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (13)
  • (2)
  • (1)
  • (13)
  • (2)
  • (1)
  • (8)
  • (16)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (13)
  • (2)
  • (4)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)