accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClPb

CAS RN 7758-95-4

IUPAC Name: λ²-lead(2+) dichloride
Synonyms: lead(II) chloride
Molecular Weight (g/mol): 278.1
Molecular Formula: Cl2Pb
InChi Key: HWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pb++]
Molecular Weight (g/mol) 
  • (22)
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (8)
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (3)
  • (3)
  • (6)
  • (2)
Grade 
  • (1)
  • (2)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (6)
  • (4)
  • (2)
  • (14)
  • (2)
  • (22)
  • (6)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (8)
  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (9)
  • (1)
  • (2)