accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7759-02-6

IUPAC Name: strontium(2+) sulfate
Synonyms: strontium sulfate
Molecular Weight (g/mol): 183.68
Molecular Formula: O4SSr
InChi Key: UBXAKNTVXQMEAG-UHFFFAOYSA-L
SMILES: [Sr++].[O-]S([O-])(=O)=O
Grade 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)