accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 776-75-0

IUPAC Name: 1-benzoylpiperidine
Synonyms: piperidine, 1-benzoyl-
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: YXTROGRGRSPWKL-UHFFFAOYSA-N
SMILES: O=C(N1CCCCC1)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)