accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBr

CAS RN 77613-65-1

IUPAC Name: 2-(bromomethyl)-1,1-difluorocyclopropane
Synonyms: 2-(bromomethyl)-1,1-difluorocyclopropane
Molecular Weight (g/mol): 170.99
Molecular Formula: C4H5BrF2
InChi Key: TUDJNSKRXIUOAJ-UHFFFAOYNA-N
SMILES: FC1(F)CC1CBr
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)