accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77628-51-4

IUPAC Name: 6-methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid
Synonyms: 6-methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: VSQMBZXCDDTZAF-UHFFFAOYSA-N
SMILES: CC1=C(N2C=CSC2=N1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)