accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONBr

CAS RN 776296-12-9

IUPAC Name: 5-bromo-3-methoxyquinoline
Synonyms: 5-bromo-3-methoxyquinoline
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: YNVIAXULFXXTLV-UHFFFAOYSA-N
SMILES: COC1=CN=C2C=CC=C(Br)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)