accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7768-28-7

IUPAC Name: 2-(2-hydroxyethyl)phenol
Synonyms: 2-(2-hydroxyethyl)phenol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: ABFCOJLLBHXNOU-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC=C1O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)