accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSNH2N

CAS RN 777-12-8

IUPAC Name: 6-(trifluoromethyl)-1,3-benzothiazol-2-amine
Synonyms: 6-(trifluoromethyl)-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 218.2
Molecular Formula: C8H5F3N2S
InChi Key: WEDYEBJLWMPPOK-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(C=C2S1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)