accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 777063-41-9

IUPAC Name: 1-(3-methylbutyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-(3-methylbutyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 264.18
Molecular Formula: C14H25BN2O2
InChi Key: YFNOUGJZNHTYAD-UHFFFAOYSA-N
SMILES: CC(C)CCN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results