accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77723-03-6

IUPAC Name: 4-methylpiperazine-1-carboximidamide hydroiodide
Synonyms: 4-methylpiperazine-1-carboximidamide hydroiodide
Molecular Weight (g/mol): 270.12
Molecular Formula: C6H15IN4
InChi Key: KESCAEZAONDSRV-UHFFFAOYSA-N
SMILES: I.CN1CCN(CC1)C(N)=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results