accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77758-84-0

IUPAC Name: phenyl(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)iodanium trifluoromethanesulfonate
Synonyms: phenyl(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)iodanium triflate
Molecular Weight (g/mol): 672.12
Molecular Formula: C13H5F16IO3S
InChi Key: GGWYKYDTNLXUMF-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)[I+]C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results