accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 77779-49-8

IUPAC Name: ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
Molecular Weight (g/mol): 253.66
Molecular Formula: C12H9ClFNO2
InChi Key: PPHSOQWURBALSQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=CC(F)=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results