accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7779-30-8

IUPAC Name: (1E)-1-(2,6,6-trimethylcyclohex-2-en-1-yl)pent-1-en-3-one
Synonyms: methylionone irone methyl α-ionone isomers
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: VPKMGDRERYMTJX-CMDGGOBGNA-N
SMILES: CCC(=O)\C=C\C1C(C)=CCCC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results