CAS RN 7779-78-4
IUPAC Name:
4-methyl-1-phenylpentan-2-ol
Synonyms:
4-methyl-1-phenyl-2-pentanol
Molecular Weight (g/mol):
178.28
Molecular Formula:
C12H18O
InChi Key:
IUADYGVMSDKSMB-UHFFFAOYNA-N
SMILES:
CC(C)CC(O)CC1=CC=CC=C1
1
–
1
of
1
results
1
4-Methyl-1-phenyl-2-pentanol 96.0+%, TCI America™
CAS: 7779-78-4 Molecular Formula: C12H18O Molecular Weight (g/mol): 178.275 MDL Number: MFCD00209515 InChI Key: IUADYGVMSDKSMB-UHFFFAOYSA-N PubChem CID: 62661 IUPAC Name: 4-methyl-1-phenylpentan-2-ol SMILES: CC(C)CC(CC1=CC=CC=C1)O