accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7782-64-1

IUPAC Name: manganese(2+) difluoride
Synonyms: manganese(II) fluoride
Molecular Weight (g/mol): 92.93
Molecular Formula: F2Mn
InChi Key: CTNMMTCXUUFYAP-UHFFFAOYSA-L
SMILES: [F-].[F-].[Mn++]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)