accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7782-92-5

IUPAC Name: sodium azanide
Synonyms: sodium amide
Molecular Weight (g/mol): 39.01
Molecular Formula: H2NNa
InChi Key: ODZPKZBBUMBTMG-UHFFFAOYSA-N
SMILES: [NH2-].[Na+]
Molecular Weight (g/mol) 
  • (5)
  • (5)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (5)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (6)