accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOHO

CAS RN 7782-99-2

IUPAC Name: sulfurous acid
Synonyms: sulfurous acid sodium bisulfite solution trioxosulfuric acid sulfur dioxide solution sodium bisulfite gr hydrogentrioxosulfate(1-) hydrogentrioxosulfate(IV) sodium bisulfite extra pure monohydrogentrioxosulfate sulfite (HSO3 1-)
Molecular Weight (g/mol): 82.07
Molecular Formula: H2O3S
InChi Key: LSNNMFCWUKXFEE-UHFFFAOYSA-N
SMILES: OS(O)=O
Grade 
  • (5)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)