accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7783-03-1

IUPAC Name: dioxotungstendiol
Synonyms: tungstic acid tungstic(VI) acid dihydrogen wolframate
Molecular Weight (g/mol): 249.85
Molecular Formula: H2O4W
InChi Key: CMPGARWFYBADJI-UHFFFAOYSA-L
SMILES: O[W](O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)