accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7783-11-1

IUPAC Name: diammonium hydrate sulfite
Synonyms: diammonium hydrate sulfite
Molecular Weight (g/mol): 134.15
Molecular Formula: H10N2O4S
InChi Key: ASPXBYAQZVXSNS-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].O.[O-]S([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results