CAS RN 7783-28-0
Ammonium phosphate, dibasic, 98+%, ACS reagent
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 SMILES: N.N.OP(O)(O)=O
Ammonium Phosphate Dibasic (Crystalline/Certified ACS), Fisher Chemical
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
Ammonium hydrogen phosphate, 98%
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: diazanium;hydrogen phosphate SMILES: N.N.OP(O)(O)=O
Ammonium phosphate, dibasic, 99+%, for analysis
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
Ammonium Phosphate, Dibasic, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
Ammonium Phosphate, Granular, NF, 96-102%, Spectrum™ Chemical
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
Ammonium phosphate, dibasic, 99%, for biochemistry
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 SMILES: N.N.OP(O)(O)=O
Ammonium Phosphate Dibasic, Crystal, ACS, 98%, Spectrum™ Chemical
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
Ammonium hydrogen phosphate, ACS, 98.0% min
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: diazanium;hydrogen phosphate SMILES: N.N.OP(O)(O)=O
di-Ammonium Hydrogen Phosphate, MilliporeSigma™
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
LiChropur™ Ammonium phosphate dibasic, 99.0-101.0% (T), MilliporeSigma™ Supelco™
MDL Number: MFCD00010891 Synonym: Ammonium hydrogenphosphate; Diammonium hydrogenphosphate; di-Ammonium hydrogenphosphate (sec)
Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 SMILES: N.N.OP(O)(O)=O
Ammonium Phosphate, Dibasic, ACS Reagent Grade, Ricca Chemical
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
Ammonium Phosphate TS/RS, 13% (w/v), Ricca Chemical
CAS: 7783-28-0 Molecular Formula: H9N2O4P Molecular Weight (g/mol): 132.06 MDL Number: MFCD00010891 InChI Key: MNNHAPBLZZVQHP-UHFFFAOYSA-N Synonym: diammonium phosphate,diammonium hydrogenphosphate,diammonium hydrogen phosphate,phosphoric acid, diammonium salt,fyrex,pelor,akoustan a,diammonium orthophosphate,diammonium acid phosphate,dibasic ammonium phosphate PubChem CID: 24540 ChEBI: CHEBI:63051 IUPAC Name: phosphoric acid diamine SMILES: N.N.OP(O)(O)=O
- 1
- 2