accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7783-48-4

IUPAC Name: strontium(2+) difluoride
Synonyms: strontium fluoride
Molecular Weight (g/mol): 125.62
Molecular Formula: F2Sr
InChi Key: FVRNDBHWWSPNOM-UHFFFAOYSA-L
SMILES: [F-].[F-].[Sr++]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)
  • (1)