accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7783-49-5

IUPAC Name: zinc(2+) difluoride
Synonyms: zinc fluoride
Molecular Weight (g/mol): 103.38
Molecular Formula: F2Zn
InChi Key: BHHYHSUAOQUXJK-UHFFFAOYSA-L
SMILES: [F-].[F-].[Zn++]
Molecular Weight (g/mol) 
  • (6)
  • (2)
Physical Form 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)