accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7783-51-9

IUPAC Name: gallium(3+) trifluoride
Synonyms: gallium(III) fluoride
Molecular Weight (g/mol): 126.72
Molecular Formula: F3Ga
InChi Key: WXXZSFJVAMRMPV-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Ga+3]
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results