accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFMn

CAS RN 7783-53-1

IUPAC Name: manganese(3+) trifluoride
Synonyms: manganese(III) fluoride
Molecular Weight (g/mol): 111.93
Molecular Formula: F3Mn
InChi Key: SRVINXWCFNHIQZ-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Mn+3]
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)