accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFSn

CAS RN 7783-62-2

IUPAC Name: tin(4+) tetrafluoride
Synonyms: tin(IV) fluoride
Molecular Weight (g/mol): 194.7
Molecular Formula: F4Sn
InChi Key: YUOWTJMRMWQJDA-UHFFFAOYSA-J
SMILES: [F-].[F-].[F-].[F-].[Sn+4]
Grade 
  • (11)
Quantity 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results