CAS RN 7783-85-9
ORP Standard, +600 mV, Ricca Chemical
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L IUPAC Name: λ2-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferrous Ammonium Sulfate, 0.00282 N (0.00282 M), 1 mL = 0.1 mg Cl2, Ricca Chemical
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L IUPAC Name: λ2-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Ferrous ammonium sulfate,Mohr's salt IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferrous Ammonium Sulfate Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Mohr Salt,Iron (II) Ammonium Sulfate Hexahydrate IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
ORP Standard, +400 mV, Ricca Chemical
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L IUPAC Name: λ2-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ammonium iron(II) sulfate hexahydrate, 99%
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Iron(II) ammonium sulfate; Ferrous ammonium sulfate IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferrous Ammonium Sulfate Hexahydrate, 98.5-101.5%, (ACS Reagent Grade), MP Biomedicals
CAS: 7783-85-9 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Ferrous ammonium sulfate,Mohr's salt IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferrous Iron Standard, 50 ppm Fe2+, Ricca Chemical
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Ammonium Iron (II) Sulfate,Iron (II) Ammonium Sulfate Hexahydrate,Iron(II) ammonium sulfate PubChem CID: 115148 IUPAC Name: λ2-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ammonium iron(II) sulfate hexahydrate, ACS, 98.5-101.5%
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Iron(II) ammonium sulfate,Ferrous ammonium sulfate IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferrous Ammonium Sulfate, Hexahydrate, Crystal, ACS, 98.5-101.5%, Spectrum™ Chemical
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L IUPAC Name: λ2-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferrous Ammonium Sulfate, 0.1N, Standardized Solution, Spectrum™ Chemical
Ferrous Ammonium Sulfate, Hexahydrate, 98.5 to 101.5%, ACS, MilliporeSigma™
CAS: 7783-85-9 Molecular Formula: FeH20N2O14S2 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L Synonym: Ferrous ammonium sulfate,Mohr's salt,Iron(II) ammonium sulfate IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Hydrogen Peroxide, 30%, Solution, Reagent, 29.0-32.0%, Spectrum™ Chemical
Ferrous Ammonium Sulfate, 6-Hydrate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7783-85-9 Molecular Weight (g/mol): 392.13 MDL Number: MFCD00150530 InChI Key: MQLVWQSVRZVNIP-UHFFFAOYSA-L IUPAC Name: λ²-iron(2+) diammonium hexahydrate disulfate SMILES: [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O