accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaAlClClClCl

CAS RN 7784-16-9

IUPAC Name: sodium tetrachloroalumanuide
Synonyms: sodium tetrachloroaluminate(1-)
Molecular Weight (g/mol): 191.77
Molecular Formula: AlCl4Na
InChi Key: CMLRNXGMTZKKSR-UHFFFAOYSA-J
SMILES: [Na+].Cl[Al-](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)