accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7784-30-7

IUPAC Name: aluminium(3+) phosphate
Synonyms: aluminium phosphate
Molecular Weight (g/mol): 121.95
Molecular Formula: AlO4P
InChi Key: ILRRQNADMUWWFW-UHFFFAOYSA-K
SMILES: [Al+3].[O-]P([O-])([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (1)