accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaOPOOPOOOPOOO

CAS RN 7785-84-4

IUPAC Name: trisodium trioxo-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-tris(olate)
Synonyms: sodium trimetaphosphate
Molecular Weight (g/mol): 305.88
Molecular Formula: Na3O9P3
InChi Key: UGTZMIPZNRIWHX-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)O1
Grade 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)