accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COPOOH3COH3C(E)

CAS RN 7786-34-7

IUPAC Name: methyl (2E)-3-[(dimethoxyphosphoryl)oxy]but-2-enoate
Synonyms: mevinphos (trans)
Molecular Weight (g/mol): 224.15
Molecular Formula: C7H13O6P
InChi Key: GEPDYQSQVLXLEU-AATRIKPKSA-N
SMILES: COC(=O)\C=C(/C)OP(=O)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results