accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7787-61-3

IUPAC Name: bismuth(3+) trifluoride
Synonyms: bismuth trifluoride
Molecular Weight (g/mol): 265.98
Molecular Formula: BiF3
InChi Key: BRCWHGIUHLWZBK-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Bi+3]
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)