accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7787-93-1

CH3SiClClCl

CAS RN 7787-93-1

IUPAC Name: dichloro(3-chloropropyl)methylsilane
Synonyms: dichloro(3-chloropropyl)methylsilane
Molecular Weight (g/mol): 191.55
Molecular Formula: C4H9Cl3Si
InChi Key: UCJHMXXKIKBHQP-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CCCCl
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)