accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7788-97-8

IUPAC Name: chromium trifluoride
Synonyms: chromium trifluoride
Molecular Weight (g/mol): 108.99
Molecular Formula: CrF3
InChi Key: QZEZRQVFRMLNPK-UHFFFAOYSA-N
SMILES: [F].[F].[F].[Cr]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results