accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4OCrOOOCrOOO

CAS RN 7789-09-5

IUPAC Name: diammonium [(oxidodioxochromio)oxy]chromiumoylolate
Synonyms: ammonium dichromate
Molecular Weight (g/mol): 252.06
Molecular Formula: Cr2H8N2O7
InChi Key: JOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (32)
  • (5)
Grade 
  • (1)
  • (4)
  • (8)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
Quantity 
  • (4)
  • (2)
  • (2)
  • (1)
  • (7)
Physical Form 
  • (3)
  • (4)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (13)
  • (8)
  • (9)
  • (2)
  • (3)
  • (32)
  • (5)
  • (1)
  • (4)
  • (8)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (7)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (8)
  • (8)
  • (1)