accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFe

CAS RN 7789-28-8

IUPAC Name: λ²-iron(2+) difluoride
Synonyms: iron(II) fluoride
Molecular Weight (g/mol): 93.84
Molecular Formula: F2Fe
InChi Key: FZGIHSNZYGFUGM-UHFFFAOYSA-L
SMILES: [F-].[F-].[Fe++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results