CAS RN 7789-48-2
IUPAC Name:
magnesium(2+) dibromide
Synonyms:
magnesium bromide
Molecular Weight (g/mol):
184.11
Molecular Formula:
Br2Mg
InChi Key:
OTCKOJUMXQWKQG-UHFFFAOYSA-L
SMILES:
[Mg++].[Br-].[Br-]