accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7789-49-3

IUPAC Name: dibromonickel trihydrate
Synonyms: dibromonickel trihydrate
Molecular Weight (g/mol): 272.55
Molecular Formula: Br2H6NiO3
InChi Key: UQPSGBZICXWIAG-UHFFFAOYSA-L
SMILES: O.O.O.Br[Ni]Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)