accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OBrBrSr

CAS RN 7789-53-9

IUPAC Name: strontium(2+) hexahydrate dibromide
Synonyms: strontium(2+) hexahydrate dibromide
Molecular Weight (g/mol): 355.52
Molecular Formula: Br2H12O6Sr
InChi Key: FLMJUJXBFKFYOZ-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Br-].[Br-].[Sr++]
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)