accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSeBrBrBr

CAS RN 7789-65-3

IUPAC Name: tetrabromo-λ⁴-selane
Synonyms: tetrabromo-λ⁴-selane
Molecular Weight (g/mol): 398.59
Molecular Formula: Br4Se
InChi Key: VTQZBGAODFEJOW-UHFFFAOYSA-N
SMILES: Br[Se](Br)(Br)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results