accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSiBrBrBr

CAS RN 7789-66-4

IUPAC Name: tetrabromosilane
Synonyms: silicon tetrabromide
Molecular Weight (g/mol): 347.7
Molecular Formula: Br4Si
InChi Key: AIFMYMZGQVTROK-UHFFFAOYSA-N
SMILES: Br[Si](Br)(Br)Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)