accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7789-67-5

IUPAC Name: tin(IV) bromide
Synonyms: tin(IV) bromide
Molecular Weight (g/mol): 442.36
Molecular Formula: Br4H4Sn
InChi Key: QENMVORAJVNRHX-UHFFFAOYSA-N
SMILES: Br.Br.Br.Br.[Sn]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results